Merck
  • Molecular Dynamics Simulation Studies of Self-Assembly of Racemic (R)/(S)-2-Bromohexadecanoic Acid on a Graphite Surface: Enantio-pure or Enantio-mixed Domains?.
Molecular Dynamics Simulation Studies of Self-Assembly of Racemic (R)/(S)-2-Bromohexadecanoic Acid on a Graphite Surface: Enantio-pure or Enantio-mixed Domains?.
The Journal of Physical Chemistry C (2007)
Ilan B, et al.