Merck
  • Solvent-free hydrothermal synthesis of anatase TiO2nanoparticleswith enhanced photocatalytic hydrogen production activity.
Solvent-free hydrothermal synthesis of anatase TiO2nanoparticleswith enhanced photocatalytic hydrogen production activity.
Applied Catalysis A: General (2013)
Ayad F. Alkaim