Merck
  • Disruption of a novel gene, DIRC3, and expression of DIRC3-HSPBAP1 fusion transcripts in a case of familial renal cell cancer and t(2;3)(q35;q21).
Disruption of a novel gene, DIRC3, and expression of DIRC3-HSPBAP1 fusion transcripts in a case of familial renal cell cancer and t(2;3)(q35;q21).
Genes, chromosomes & cancer (2003-08-27)
Daniëlle Bodmer, Marga Schepens, Marc J Eleveld, Eric F P M Schoenmakers, Ad Geurts van Kessel
ZUSAMMENFASSUNG
Previously, we identified a family with renal cell cancer and a t(2;3)(q35;q21). Positional cloning of the chromosome 3 breakpoint led to the identification of a novel gene, DIRC2, that spans this breakpoint. Here we have characterized the chromosome 2 breakpoint in detail and found that another novel gene, designated DIRC3, spans this breakpoint. In addition, we found that the first two exons of DIRC3 can splice to the second exon of HSPBAP1, a JmjC-Hsp27 domain gene that maps proximal to the breakpoint on chromosome 3. This splice results in the formation of DIRC3-HSPBAP1 fusion transcripts. We propose that these fusion transcripts may affect normal HSPBAP1 function and concomitant chromatin remodeling and/or stress response signals within t(2;3)(q35;q21)-positive kidney cells. As a consequence, familial renal cell cancer may develop.