Merck
  • Structure determination of the Diels-Alder product of a ketovinylphosphonate with< i> E</i>-1-acetoxy-1, 3-butadiene.
Structure determination of the Diels-Alder product of a ketovinylphosphonate with&lt; i&gt; E&lt;/i&gt;-1-acetoxy-1, 3-butadiene.
Tetrahedron Letters (1996)
McClure CK, et al.