Merck
  • A novel, evolutionarily conserved gene family with putative sequence-specific single-stranded DNA-binding activity.
A novel, evolutionarily conserved gene family with putative sequence-specific single-stranded DNA-binding activity.
Genomics (2002-06-25)
Patricia Castro, Hong Liang, Jan C Liang, Lalitha Nagarajan
ZUSAMMENFASSUNG
Complete and partial deletions of chromosome 5q are recurrent cytogenetic anomalies associated with aggressive myeloid malignancies. Earlier, we identified an approximately 1.5-Mb region of loss at 5q13.3 between the loci D5S672 and D5S620 in primary leukemic blasts. A leukemic cell line, ML3, is diploid for all of chromosome 5, except for an inversion-coupled translocation within the D5S672-D5S620 interval. Here, we report the development of a bacterial artificial chromosome (BAC) contig to define the breakpoint and the identification of a novel gene SSBP2, the target of disruption in ML3 cells. A preliminary evaluation of SSBP2 as a tumor suppressor gene in primary leukemic blasts and cell lines suggests that the remaining allele does not undergo intragenic mutations. SSBP2 is one of three members of a closely related, evolutionarily conserved, and ubiquitously expressed gene family. SSBP3 is the human ortholog of a chicken gene, CSDP, that encodes a sequence-specific single-stranded DNA-binding protein. SSBP3 localizes to chromosome 1p31.3, and the third member, SSBP4, maps to chromosome 19p13.1. Chromosomal localization and the putative single-stranded DNA-binding activity suggest that all three members of this family are capable of potential tumor suppressor activity by gene dosage or other epigenetic mechanisms.