Merck
  • Characterization of Lactoferrin (LF) from Colostral Whey Using Anti-LF Antibody Immunoaffinity Chromatography
Characterization of Lactoferrin (LF) from Colostral Whey Using Anti-LF Antibody Immunoaffinity Chromatography
Journal of Food Science (2002)
Y.-Y. Tu, C.-C. Chen, J-H. Chang and H-M. Chang.