Merck
The PX domains of p47phox and p40phox bind to lipid products of PI(3)K.
Nature cell biology (2001-07-04)
F Kanai, H Liu, S J Field, H Akbary, T Matsuo, G E Brown, L C Cantley, M B Yaffe
ZUSAMMENFASSUNG
PX domains are found in a variety of proteins that associate with cell membranes, but their molecular function has remained obscure. We show here that the PX domains in p47phox and p40phox subunits of the phagocyte NADPH oxidase bind to phosphatidylinositol-3,4-bisphosphate (PtdIns(3,4)P(2)) and phosphatidylinositol-3-phosphate (PtdIns(3)P), respectively. We also show that an Arg-to-Gln mutation in the PX domain of p47phox, which is found in patients with chronic granulomatous disease, eliminates phosphoinositide binding, as does the analogous mutation in the PX domain of p40phox. The PX domain of p40phox localizes specifically to PtdIns(3)P-enriched early endosomes, and this localization is disrupted by inhibition of phosphoinositide-3-OH kinase (PI(3)K) or by the Arg-to-Gln point mutation. These findings provide a molecular foundation to understand the role of PI(3)K in regulating neutrophil function and inflammation, and to identify PX domains as specific phosphoinositide-binding modules involved in signal transduction events in eukaryotic cells.