Merck
  • t-BuOK-Catalyzed addition of ketones and nitriles to vinylic silanes, phosphines and thio derivatives.
<I>t-</I>BuOK-Catalyzed addition of ketones and nitriles to vinylic silanes, phosphines and thio derivatives.
Chemical Communications (Cambridge, England) (2001)
Bunlaksananusorn T, et al.