Merck
  • Synthesis of indole, benzofuran and benzothiophene by reaction of 2-anilinoethanol, 2-phenoxyethanol and 2-(phenylthio) ethanol on AlPO4 and Pd/AlPO4 catalysts.
Synthesis of indole, benzofuran and benzothiophene by reaction of 2-anilinoethanol, 2-phenoxyethanol and 2-(phenylthio) ethanol on AlPO<SUB>4</SUB> and Pd/AlPO<SUB>4</SUB> catalysts.
J. Mol. Catal. A: Chem. (1995)
Afxantidis J, et al.