- Identity-by-descent filtering of exome sequence data identifies PIGV mutations in hyperphosphatasia mental retardation syndrome.
Identity-by-descent filtering of exome sequence data identifies PIGV mutations in hyperphosphatasia mental retardation syndrome.
Nature genetics (2010-08-31)
Peter M Krawitz, Michal R Schweiger, Christian Rödelsperger, Carlo Marcelis, Uwe Kölsch, Christian Meisel, Friederike Stephani, Taroh Kinoshita, Yoshiko Murakami, Sebastian Bauer, Melanie Isau, Axel Fischer, Andreas Dahl, Martin Kerick, Jochen Hecht, Sebastian Köhler, Marten Jäger, Johannes Grünhagen, Birgit Jonske de Condor, Sandra Doelken, Han G Brunner, Peter Meinecke, Eberhard Passarge, Miles D Thompson, David E Cole, Denise Horn, Tony Roscioli, Stefan Mundlos, Peter N Robinson
PMID20802478
ZUSAMMENFASSUNG
Hyperphosphatasia mental retardation (HPMR) syndrome is an autosomal recessive form of mental retardation with distinct facial features and elevated serum alkaline phosphatase. We performed whole-exome sequencing in three siblings of a nonconsanguineous union with HPMR and performed computational inference of regions identical by descent in all siblings to establish PIGV, encoding a member of the GPI-anchor biosynthesis pathway, as the gene mutated in HPMR. We identified homozygous or compound heterozygous mutations in PIGV in three additional families.