Merck
Identification of a novel small-molecule agonist for human G protein-coupled receptor 3.
The Journal of pharmacology and experimental therapeutics (2014-03-19)
Chenli Ye, Zhenghong Zhang, Zhilong Wang, Qiuhong Hua, Ru Zhang, Xin Xie
ZUSAMMENFASSUNG
G protein-coupled receptor 3 (GPR3) is an orphan G protein-coupled receptor (GPCR) predominantly expressed in mammalian brain and oocytes. GPR3 plays important roles in these two organs and is known as a Gαs-coupled receptor-activated constitutively in cells. However, the signal transduction pathway and pharmacological function of GPR3 remain unclear because of the lack of a specific ligand. By use of a human embryonic kidney 293 cell line stably expressing FLAG-GPR3-green fluorescent protein, a chemical screening for GPR3 ligands was performed using homogeneous time-resolved fluorescence cAMP assay. Diphenyleneiodonium chloride (DPI) was identified as a novel agonist of GPR3 with weak or no cross-reactivity with other GPCRs. DPI was further characterized to activate several GPR3-mediated signal transduction pathways, including Ca(2+) mobilization, cAMP accumulation, membrane recruitment of β-arrestin2, and receptor desensitization. Parallel studies revealed that the activity of DPI is much more pronounced than sphingosine 1-phosphate, a previously reported GPR3 agonist. Our study identified a novel and specific agonist of GPR3, which provides a useful tool for further study of this orphan GPCR.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Diphenyleniodoniumchlorid, ≥98%