Merck
  • Synthesis of ?-Keto Acids and Methyl Ketones Using Bis (trimethylsilyl) Malonate and Triethylamine in the Presence of Lithium or Magnesium Halides.
Synthesis of ?-Keto Acids and Methyl Ketones Using Bis (trimethylsilyl) Malonate and Triethylamine in the Presence of Lithium or Magnesium Halides.
Synthetic Communications (1985)
Rathke MW and Nowak MA.