- Facile synthesis of oxabicyclic alkenes by ultrasonication-promoted diels-alder cycloaddition of furano dienes.
Facile synthesis of oxabicyclic alkenes by ultrasonication-promoted diels-alder cycloaddition of furano dienes.
The Journal of organic chemistry (2004-08-17)
Kun Wei, Hai-Tao Gao, Wei-Dong Z Li
PMID15307754
ZUSAMMENFASSUNG
Ultrasonic irradiation effectively promotes the Diels-Alder reaction of substituted furans with reactive dienophiles (i.e., dimethyl acetylenedicarboxylate (DMAD) and dimethyl maleate). Regiospecific furano Diels-Alder cycloaddition of 2-vinylic furans with DMAD furnished functionalized oxabicyclic alkenes in good yield under ultrasonication condition.
MATERIALIEN