Merck
Mutation in FAM134B causing severe hereditary sensory neuropathy.
Journal of neurology, neurosurgery, and psychiatry (2010-12-01)
Sinéad M Murphy, Gabrielle L Davidson, Sebastian Brandner, Henry Houlden, Mary M Reilly