Merck
  • Polycyclic fluoroaromatic compounds. Part XII.[1] The extent of 1-substitution in octafluoronaphthalene.
Polycyclic fluoroaromatic compounds. Part XII.[1] The extent of 1-substitution in octafluoronaphthalene.
Journal of Fluorine Chemistry (1985)
Burdon J and Rimmington TW.