Merck
PET imaging of adrenal cortical tumors with the 11beta-hydroxylase tracer 11C-metomidate.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine (2000-11-18)
M D Gross, B Shapiro, R T Kloos, B Dwamena
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Etomidat-Unreinheit B, European Pharmacopoeia (EP) Reference Standard