Merck
  • Micellar electrokinetic and high-performance liquid chromatographic determination of potential manufacturing impurities in pholcodine.
Micellar electrokinetic and high-performance liquid chromatographic determination of potential manufacturing impurities in pholcodine.
Journal of chromatography. A (2000-12-06)
O M Denk, D G Watson, G G Skellern
ZUSAMMENFASSUNG
Quantitative high-performance liquid chromatographic (HPLC) and micellar electrokinetic chromatographic (MEKC) methods have been developed for the determination of four structurally related potential manufacturing impurities, including morphine, of the opiate derivative pholcodine. Pholcodine and the four impurities were separated by MEKC in less than 14 min using a 70 cm x 75 microm I.D. uncoated fused-silica capillary (25 kV at 30 degrees C) and a running buffer consisting of 10% acetonitrile (v/v) in 20 mM borate-phosphate buffer pH 8.0 containing 40 mM sodium dodecyl sulphate (SDS). The MEKC method was compared to a HPLC method using a 5 microm Luna phenyl-hexyl column (150 x 4.6 mm I.D.) eluted with a mobile phase consisting of a mixture of 10% (v/v) acetonitrile, 7% (v/v) tetrahydrofuran in 20 mM phosphate buffer pH 8.0. Both methods were fully validated and a comparison was made regarding selectivity, linearity, precision, robustness and limits of detection and quantitation. The presence of the impurities in different samples of pholcodine drug substance was investigated using both methods.