- Etilefrine for the prevention of priapism in adult sickle cell disease.
Etilefrine for the prevention of priapism in adult sickle cell disease.
British journal of haematology (2002-08-16)
Iheanyi Okpala, Neill Westerdale, Tina Jegede, Betty Cheung
PMID12181066
ZUSAMMENFASSUNG
Priapism is a common complication of sickle cell disease (SCD) that could lead to erectile dysfunction and psychosocial problems. Treatment of established fulminant priapism is usually not satisfactory. It is therefore important to prevent this complication of SCD. The alpha-adrenergic agonist etilefrine (50-100 mg/d) produced a good clinical response in 13 of 18 (72%) adults who have recurrent priapism; 17 had SCD and one sickle cell trait. After a follow-up of 1-48 months, none of the 18 people on etilefrine developed hypertension or sexual dysfunction. Similar efficacy and safety profiles of the drug have been reported previously.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Etilefrin -hydrochlorid, European Pharmacopoeia (EP) Reference Standard