Merck
Morierinin: a new cytotoxic cucurbitacin from the leaves of Morierina montana Vieill.
Natural product research (2003-06-26)
Marc Litaudon, Christianne Gaspard, Thierry Sévenet
ZUSAMMENFASSUNG
An extract of the leaves of Morierina montana collected in New Caledonia was found to be cytotoxic for KB cells. A bioassay-guided fractionation procedure led to the isolation of a series of cucurbitacins of which one, morierinin (1), is new. The isolation and structure elucidation of 1 are described.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cucurbitacin E, ≥95% (HPLC)