Merck
Pharmacokinetics of pramiracetam in healthy volunteers after oral administration.
International journal of clinical pharmacology research (1992-01-01)
A Auteri, P Blardi, G Celasco, G Segre, R Urso
ZUSAMMENFASSUNG
The pharmacokinetics of pramiracetam was assessed using an HPLC method after oral administration of two different formulations of 600 mg (a solution and a tablet) of pramiracetam to 11 fasting volunteers. The mean kinetic parameters were: t1 = 4.7 +/- 2.4 - 4.3 +/- 2.2 h, AUC = 57.6 +/- 43.6 - 47.2 +/- 33.9 micrograms h/ml, Cmax = 6.80 +/- 3.2 - 5.80 +/- 3.3 micrograms/ml for the solution and the tablet respectively. The plasma profile of pramiracetam proved to be not highly affected by the formulation, only that the absorption rate was faster after oral administration of the drug in solution than after administration as a tablet. The half-life was very variable between subjects [2-8 hours], but less variable within subjects and it was unaffected by the formulation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Pramiracetam, ≥98% (HPLC)