Merck
Cyanogenic glucosides in grapevine: polymorphism, identification and developmental patterns.
Phytochemistry (2005-01-18)
T K Franks, Y Hayasaka, S Choimes, R van Heeswijck
ZUSAMMENFASSUNG
Twelve grapevine (Vitis vinifera L.) cultivars were surveyed for 'cyanide potential' (i.e. the total cyanide measured in beta-glucosidase-treated crude, boiled tissue extract) in mature leaves. Two related cultivars (Carignan and Ruby Cabernet) had mean cyanide potential (equivalent to 110 mgHCNkg-1fr.wt) ca. 25-fold greater than that of the other 10 cultivars, and so the trait is polymorphic in the species. In boiled leaf extracts of Carignan and Ruby Cabernet, free cyanide constituted a negligible fraction of the total cyanide potential because beta-glucosidase treatment was required to liberate the major cyanide fraction - which is therefore bound in glucosylated cyanogenic compound(s) (or cyanogenic glucosides). In addition, cyanide was liberated from ground leaf tissue of Ruby Cabernet but not Sultana (a cultivar with low cyanide potential). Hence, the high cyanide potential in Ruby Cabernet leaves is coupled with endogenous beta-glucosidase(s) activity and this cultivar may be considered 'cyanogenic'. A method was developed to detect and identify cyanogenic glucosides using liquid chromatography combined with tandem mass spectrometry (LC-MS/MS). Two putative cyanogenic glucosides were found in extracts from leaves of Carignan and Ruby Cabernet and were identified as the epimers prunasin and sambunigrin. Cyanide potential measured at three times over the growing season in young and mature leaves, petioles, tendrils, flowers, berries, seeds and roots of Ruby Cabernet was substantially higher in the leaves compared with all other tissues. This characterisation of cyanogenic glucoside accumulation in grapevine provides a basis for gauging the involvement of the trait in interactions of the species with its pests and pathogens.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Prunasin