- Inhibition of the metabolism of brotizolam by erythromycin in humans: in vivo evidence for the involvement of CYP3A4 in brotizolam metabolism.
Inhibition of the metabolism of brotizolam by erythromycin in humans: in vivo evidence for the involvement of CYP3A4 in brotizolam metabolism.
British journal of clinical pharmacology (2005-07-27)
Takaki Tokairin, Takashi Fukasawa, Norio Yasui-Furukori, Toshiaki Aoshima, Akihito Suzuki, Yoshimasa Inoue, Tomonori Tateishi, Koichi Otani
PMID16042670
ZUSAMMENFASSUNG
To obtain in vivo evidence for the involvement of cytochrome P450 (CYP) 3A4 in the metabolism of brotizolam. Fourteen healthy male volunteers received erythromycin 1200 mg day(-1) or placebo for 7 days in a double-blind randomized crossover manner. On the 6th day they received a single oral 0.5-mg dose of brotizolam, and blood samplings were performed for 24 h. Erythromycin treatment significantly increased the peak plasma concentration (P < 0.05), total area under the plasma concentration-time curve (P < 0.01), and elimination half-life (P < 0.01) of brotizolam. The present study provides in vivo evidence for the involvement of CYP3A4 in brotizolam metabolism.