Merck
[Studies on chemical constituents in roots of Heracleum rapula].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2006-05-19)
Yuan-Yan Liu, Cun Zhang, Li Li, Yong-Qing Xiao
ZUSAMMENFASSUNG
To study the chemical constituents in roots of Heracleum rapula. The constituents were isolated by column chromatography on silica gel and ODS, and identified by spectroscopic methods. Nine compounds, osthol ( I ), bergapten (II), xanthotoxin (III), isopimpinellin (IV), imperatorin (V), isoimperatorin (VI), cnidilin (VII), phellopterin (VIII), rivulobirin A (IX) were isolated and identified. Compounds VI, VII, VIII, IX were isolated from this plant for the first time.