Merck
  • Inhibition of human thiopurine S-methyltransferase by various nonsteroidal anti-inflammatory drugs in vitro: a mechanism for possible drug interactions.
Inhibition of human thiopurine S-methyltransferase by various nonsteroidal anti-inflammatory drugs in vitro: a mechanism for possible drug interactions.
Drug metabolism and disposition: the biological fate of chemicals (2007-06-08)
Kersti Oselin, Kaili Anier
ZUSAMMENFASSUNG
Thiopurine S-methyltransferase (TPMT) is a biotransformation phase II enzyme responsible for the metabolic inactivation of thiopurine drugs. The present study was carried out to investigate the inhibitory potential of 15 nonsteroidal anti-inflammatory drugs (NSAIDs) on human TPMT activity in vitro. TPMT activity was measured in pooled human erythrocytes in the absence and presence of various NSAIDs using the previously published high-performance liquid chromatography-UV method. To determine the inhibition type and K(i) value for each compound, we performed kinetic analysis at five different inhibitor concentrations close to the IC(50) value obtained in preliminary experiments. Naproxen (K(i) = 52 microM), mefenamic acid (K(i) = 39 microM), and tolfenamic acid (K(i) = 50 microM) inhibited TPMT activity in a noncompetitive manner. The estimated K(i) values for the inhibition of TPMT by ketoprofen (K(i) = 172 microM) and ibuprofen (K(i) = 1043 microM) indicated that the propionic acid derivatives were relatively weak inhibitors of TPMT. Our results suggest that coadministration of thiopurines and various NSAIDs may lead to drug interactions.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Olsalazin-Natrium, European Pharmacopoeia (EP) Reference Standard
Olsalazin-Natrium für Leistungstest, European Pharmacopoeia (EP) Reference Standard