Merck
Steroid-sparing effect of pimecrolimus cream 1% in children with severe atopic dermatitis.
Dermatology (Basel, Switzerland) (2007-10-04)
Torsten Zuberbier, Lucie Heinzerling, Thomas Bieber, Uwe Schauer, Sven Klebs, Matthias Bräutigam
ZUSAMMENFASSUNG
To investigate if pimecrolimus cream 1% reduces the need for steroids in the long-term management of severe pediatric atopic dermatitis (AD). A total of 184 pediatric patients (aged 2-17 years) with a history of severe AD according to Rajka and Langeland were enrolled. Patients were randomized to treatment with pimecrolimus cream or vehicle cream for a 24-week period. Prednicarbate 0.25% was applied as rescue medication. Patients on pimecrolimus required steroids on a mean of 29% of study days, compared with 35% of patients on vehicle (p = 0.1841). On the head and neck only, the respective figures were 10 versus 19% (p = 0.0009). In patients enrolled with acute severe disease (Investigator's Global Assessment > or = 4), steroids were used on 28% of the days in the pimecrolimus group compared to 45% in the control group (p = 0.0024). On the head and neck, steroids were used on 10% of study days with pimecrolimus versus 30% with vehicle (p < 0.0001). The results indicate that the need for topical steroids on the head and neck is reduced with pimecrolimus cream 1% in the management of severe pediatric AD according to the definition of Rajka and Langeland.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Prednicarbat für die Systemeignung A, European Pharmacopoeia (EP) Reference Standard
Prednicarbat, European Pharmacopoeia (EP) Reference Standard