Merck
  • Investigation of the anti-inflammatory potential of Aloe vera gel (97.5%) in the ultraviolet erythema test.
Investigation of the anti-inflammatory potential of Aloe vera gel (97.5%) in the ultraviolet erythema test.
Skin pharmacology and physiology (2008-02-07)
J Reuter, A Jocher, J Stump, B Grossjohann, G Franke, C M Schempp
ZUSAMMENFASSUNG
Aloe vera is a natural product that is frequently used in soothing skin care products such as aftersun lotions. In the present study we aimed to explore the anti-inflammatory potential of a highly concentrated A. vera gel in the UV erythema test in vivo. 40 volunteers with skin types II and III were included in the randomized, double-blind, placebo-controlled, phase III monocenter study. Test areas on the back were irradiated with the 1.5-fold minimal erythema dose of UVB. Subsequently, the test areas were treated occlusively on 2 subsequent days with A. vera gel (97.5%), the positive controls (0.25% prednicarbate, 1% hydrocortisone in placebo gel and 1% hydrocortisone cream) and a placebo gel. Erythema values were determined photometrically after 24 and 48 h. A. vera gel (97.5%) significantly reduced UV-induced erythema after 48 h, being superior to 1% hydrocortisone in placebo gel. In contrast, 1% hydrocortisone in cream was more efficient than A. vera gel. In this study after 48 h the A. vera gel (97.5%) displayed some anti-inflammatory effects superior to those of 1% hydrocortisone in placebo gel. The A. vera gel tested here might be useful in the topical treatment of inflammatory skin conditions such as UV-induced erythema.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Prednicarbat für die Systemeignung A, European Pharmacopoeia (EP) Reference Standard
Prednicarbat, European Pharmacopoeia (EP) Reference Standard