- [Poisonings with anticoagulant rodenticides].
[Poisonings with anticoagulant rodenticides].
Archiwum medycyny sadowej i kryminologii (2008-04-25)
Marcin Zawadzki, Jan Magdalan, Artur Niedźwiedź
PMID18432144
ZUSAMMENFASSUNG
Anticoagulant rodenticides are commonly used in extermination of rodents. Failure to adhere to safety principles and sometimes a deliberate use of these compounds may be a reason of severe poisonings in human. Diagnosis is based on a characteristic clinical course of the disease and postmortem examinations.