Merck
Anthraquinones from the roots of Prismatomeris malayana.
Natural product research (2008-07-17)
Pittaya Tuntiwachwuttikul, Yuttapon Butsuri, Piyanut Sukkoet, Uma Prawat, Walter Charles Taylor
ZUSAMMENFASSUNG
A novel anthraquinone, 1,3-dihydroxy-5,6-dimethoxy-2-methoxymethyl-9,10-anthraquinone (9) and a new natural product, 2-hydroxymethyl-1-methoxy-9,10-anthraquinone (8) were isolated from the roots of Prismatomeris malayana together with seven known anthraquinones, tectoquinone (1), 1-hydroxy-2-methyl-9,10-anthraquinone (2), rubiadin (3), rubiadin-1-methyl ether (4), 1,3-dihydroxy-5,6-dimethoxy-2-methyl-9,10-anthraquinone (5), nordamnacanthal (6), and damnacanthal (7). Their structures were determined on the basis of spectroscopic data. Some of the anthraquinones were tested for anticancer, antifungal, and antimalarial activities.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Rubiadin, analytical standard