Merck
  • Interactions of 1-methyl-1,2,3,4-tetrahydroisoquinoline with lamotrigine, oxcarbazepine, pregabalin, and topiramate in the mouse maximal electroshock-induced seizure model: a type I isobolographic analysis.
Interactions of 1-methyl-1,2,3,4-tetrahydroisoquinoline with lamotrigine, oxcarbazepine, pregabalin, and topiramate in the mouse maximal electroshock-induced seizure model: a type I isobolographic analysis.
Epilepsy research (2010-02-02)
Jarogniew J Luszczki, Lucyna Antkiewicz-Michaluk, Grzegorz Raszewski, Stanislaw J Czuczwar
ZUSAMMENFASSUNG
The aim of this study was to characterize the anticonvulsant effects of 1-methyl-1,2,3,4-tetrahydroisoquinoline (MeTHIQ--an endogenous parkinsonism-preventing substance) in combination with four second-generation antiepileptic drugs (AEDs: lamotrigine [LTG], oxcarbazepine [OXC], pregabalin [PGB], and topiramate [TPM]) in the mouse maximal electroshock (MES)-induced seizure model by using the type I isobolographic analysis for parallel and non-parallel dose-response relationship curves (DRRCs). Potential adverse-effect profiles of interactions of MeTHIQ with LTG, OXC, PGB and TPM at the fixed-ratio of 1:1 from the MES test with respect to motor performance, long-term memory and skeletal muscular strength were measured along with total brain concentrations of MeTHIQ and TPM. In the mouse MES model, MeTHIQ administered singly had its DRRC parallel to those for OXC and TPM, and simultaneously, non-parallel to those for LTG and PGB. With type I isobolography for parallel DRRCs, the combination of MeTHIQ with TPM at three fixed-ratios of 1:3, 1:1 and 3:1 exerted supra-additive (synergistic) interaction, whereas the combination of MeTHIQ with OXC at the fixed-ratios of 1:3, 1:1 and 3:1 produced additive interaction. Similarly, the type I isobolography for non-parallel DRRCs revealed that the combination of MeTHIQ with LTG and PGB at the fixed-ratio of 1:1 produced additive interaction. For all combinations, neither motor coordination, long-term memory nor muscular strength were affected. Total brain concentrations of MeTHIQ and TPM revealed no significant changes in their concentrations when the drugs were combined at the fixed-ratios of 1:3, 1:1 and 3:1. In conclusion, the synergistic interaction of MeTHIQ with TPM at the fixed-ratios of 1:3, 1:1 and 3:1 against MES-induced seizures was pharmacodynamic in nature and thus, it is worthy of consideration in further clinical settings. The combinations of MeTHIQ with LTG, OXC and PGB were neutral in the mouse MES model.