Merck
  • Anti-inflammatory activities of cucurbitacin E isolated from Citrullus lanatus var. citroides: role of reactive nitrogen species and cyclooxygenase enzyme inhibition.
Anti-inflammatory activities of cucurbitacin E isolated from Citrullus lanatus var. citroides: role of reactive nitrogen species and cyclooxygenase enzyme inhibition.
Fitoterapia (2011-08-30)
Siddig Ibrahim Abdelwahab, Loiy Elsir Ahmed Hassan, Hasnah Mohd Sirat, Sakina M Ahmed Yagi, Waleed S Koko, Syam Mohan, Manal Mohamed Elhassan Taha, Syahida Ahmad, Cheah Shiau Chuen, Putri Narrima, Mohd Mustafa Rais, A Hamid A Hadi
ZUSAMMENFASSUNG
The in vivo and in vitro mechanistic anti-inflammatory actions of cucurbitacin E (CE) (Citrullus lanatus var. citroides) were examined. The results showed that LPS/INF-γ increased NO production in RAW264.7 macrophages, whereas L-NAME and CE curtailed it. CE did not reveal any cytotoxicity on RAW264.7 and WRL-68 cells. CE inhibited both COX enzymes with more selectivity toward COX-2. Intraperitoneal injection of CE significantly suppressed carrageenan-induced rat's paw edema. ORAC and FRAP assays showed that CE is not a potent ROS scavenger. It could be concluded that CE is potentially useful in treating inflammation through the inhibition of COX and RNS but not ROS.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cucurbitacin E, ≥95% (HPLC)