- Dehydration of detomidine hydrochloride monohydrate.
Dehydration of detomidine hydrochloride monohydrate.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences (2011-08-30)
K Veldre, A Actiņš, J Jaunbergs
PMID21871958
ZUSAMMENFASSUNG
The thermodynamic stability of detomidine hydrochloride monohydrate has been evaluated on the basis of phase transition kinetics in solid state. A method free of empirical models was used for the treatment of kinetic data, and compared to several known solid state kinetic data processing methods. Phase transitions were monitored by powder X-ray diffraction (PXRD) and thermal analysis. Full PXRD profiles were used for determining the phase content instead of single reflex intensity measurements, in order to minimize the influence of particle texture. We compared the applicability of isothermal and nonisothermal methods to our investigation of detomidine hydrochlorine monohydrate dehydration.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Detomidin -hydrochlorid, European Pharmacopoeia (EP) Reference Standard