Merck
  • Positive impact of the β-blocker celiprolol on panic, anxiety, and cardiovascular parameters in patients with mitral valve prolapse syndrome.
Positive impact of the β-blocker celiprolol on panic, anxiety, and cardiovascular parameters in patients with mitral valve prolapse syndrome.
Journal of clinical psychopharmacology (2011-11-05)
Silke Bachmann, Ursula Muller-Werdan, Michael Huber, Markus Kasel, Karl Werdan, Hendrik Schmidt