Merck
  • Determination of fenticonazole enantiomers by LC-ESI-MS/MS and its application to pharmacokinetic studies in female rats.
Determination of fenticonazole enantiomers by LC-ESI-MS/MS and its application to pharmacokinetic studies in female rats.
Arzneimittel-Forschung (2011-12-15)
Zhenbin Feng, Qiaogen Zou, Xiaoheng Tan, Wenjun Che, Zunjian Zhang
ZUSAMMENFASSUNG
A simple, rapid, and specific high-performance liquid chromatograph coupled with a tandem mass spectrometry method has been developed and validated for the determination of fenticonazole (CAS 72479-26-6) enantiomers in rat plasma. Simple protein precipitation by acetonitrile was utilized for extracting analytes from the plasma samples. Chromatography separation was performed on a C18 analytical column (150 mm x 2.0 mm, 5 microm) with a mobile phase consisting of methanol-10 mM aqueous ammonium acetate (adjusted to pH 3.5 with acetic acid) (90:10, v/v) at a flow rate of 0.2 ml/min. Detection was carried out on a triple quadrupole mass spectrometer equipped with electrospray ionization (ESI) source, and operated in multiple-reaction monitoring (MRM) mode. The calibration curves were linear over the range 0.5 -200 ng/ml (r > 0.99). The relative recoveries of R-(-)-fenticonazole and its enantiomer were better than 85%. The intra- and inter-day precisions (R.S.D.%) and deviations of the assay accuracies were less than 10%. This newly developed and validated method was successfully applied to pharmacokinetic studies after administration at a single dose of 20 mg/ kg R-(-)-fenticonazole nitrate and its enantiomer to female rats per vagina. The Cmax value of S-(+)-fenticonazole was greater than that of R-(-)-fenticonazole by 1.36-fold, whereas, the t(1/2) beta and MRT values of R-(-)-fenticonazole were longer than those of its enantiomer by 1.95- and 1.24-fold. The results indicated that S-(+)-fenticonazole was faster in absorption and elimination in female rat. But, the Tmax and AUC(0-12) values for each of fenticonazole enantiomers were not significantly different.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Fenticonazolnitrat, European Pharmacopoeia (EP) Reference Standard