Merck
Arctigenin alleviates ER stress via activating AMPK.
Acta pharmacologica Sinica (2012-06-19)
Yuan Gu, Xiao-xiao Sun, Ji-ming Ye, Li He, Shou-sheng Yan, Hao-hao Zhang, Li-hong Hu, Jun-ying Yuan, Qiang Yu
ZUSAMMENFASSUNG
To investigate the protective effects of arctigenin (ATG), a phenylpropanoid dibenzylbutyrolactone lignan from Arctium lappa L (Compositae), against ER stress in vitro and the underlying mechanisms. A cell-based screening assay for ER stress regulators was established. Cell viability was measured using MTT assay. PCR and Western blotting were used to analyze gene and protein expression. Silencing of the CaMKKβ, LKB1, and AMPKα1 genes was achieved by RNA interference (RNAi). An ATP bioluminescent assay kit was employed to measure the intracellular ATP levels. ATG (2.5, 5 and 10 μmol/L) inhibited cell death and unfolded protein response (UPR) in a concentration-dependent manner in cells treated with the ER stress inducer brefeldin A (100 nmol/L). ATG (1, 5 and 10 μmol/L) significantly attenuated protein synthesis in cells through inhibiting mTOR-p70S6K signaling and eEF2 activity, which were partially reversed by silencing AMPKα1 with RNAi. ATG (1-50 μmol/L) reduced intracellular ATP level and activated AMPK through inhibiting complex I-mediated respiration. Pretreatment of cells with the AMPK inhibitor compound C (25 μmol/L) rescued the inhibitory effects of ATG on ER stress. Furthermore, ATG (2.5 and 5 μmol/L) efficiently activated AMPK and reduced the ER stress and cell death induced by palmitate (2 mmol/L) in INS-1 β cells. ATG is an effective ER stress alleviator, which protects cells against ER stress through activating AMPK, thus attenuating protein translation and reducing ER load.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Brefeldin A aus Penicillium brefeldianum, from Penicillium brefeldianum, ≥99% (HPLC and TLC)
Sigma-Aldrich
Dorsomorphin, ≥98% (HPLC)
Sigma-Aldrich
AICAR, ≥98% (HPLC), powder
Sigma-Aldrich
(−)-Arctigenin