Merck
  • Cost-effectiveness of iso- versus low-osmolality contrast media in outpatients with high risk of contrast medium-induced nephropathy.
Cost-effectiveness of iso- versus low-osmolality contrast media in outpatients with high risk of contrast medium-induced nephropathy.
Biomedica : revista del Instituto Nacional de Salud (2012-12-18)
Liliana Alejandra Chicaíza-Becerra, Mario García-Molina, Óscar Gamboa
ZUSAMMENFASSUNG
Contrast media can cause acute renal failure by direct toxic effects on the tubular cells and kidney ischemia. Diabetics and hospitalized patients have a greater risk of developing contrast-induced nephropathy than the general population. The cost effectiveness of iso and low-osmolality contrast media was assessed in high risk outpatients. The analysis was based on a systematic literature review comparing the nephrotoxic effects of iso- to low-osmolality contrast media. Only direct costs were considered; these were obtained from the official tariff manual. Incremental cost-effectiveness ratios, efficiency curves and acceptability curves were calculated. Univariate sensitivity analyses were performed for costs and effects, as well as probabilistic analyses. Zero and 3% discounts were applied to results. The cost-effectiveness threshold was equal to the per capita GDP per life-year gained. Alternatives with Iopamidol and Iodixanol are preferable to the others, because both reduce risk of contrast-induced nephropathy and are less costly. The incremental cost-effectiveness of the Iodixanol alternative compared to the Iopamidol alternative is US$ 14,660 per additional life year gained; this is more than twice the threshold. The low-osmolality contrast medium, Iopamidol, appears to be cost-effective when compared with Iohexol or other low-osmolality contrast media (Iopromide, Iobitridol, Iomeprol, Iopentol and Ioxilan) in contrast-induced nephropathy, high-risk outpatients. The choice of the iso-osmolality contrast medium, Iodixanol, depends on its cost per vial and on the willingness to pay.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
USP
Iodixanol-verwandte Verbindung D, United States Pharmacopeia (USP) Reference Standard
Iopentol, European Pharmacopoeia (EP) Reference Standard