Merck
Predictors of adherence to inhaled medications.
Journal of general internal medicine (2013-03-05)
Ami Schattner
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Ipratropiumbromid-Unreinheit B, European Pharmacopoeia (EP) Reference Standard