Merck
  • Characterization of impurities in tylosin using dual liquid chromatography combined with ion trap mass spectrometry.
Characterization of impurities in tylosin using dual liquid chromatography combined with ion trap mass spectrometry.
Talanta (2013-04-20)
Shruti Chopra, Ann Van Schepdael, Jos Hoogmartens, Erwin Adams
ZUSAMMENFASSUNG
Investigation of unknown impurities in a tylosin sample was performed using liquid chromatography coupled to mass spectrometry (LC/MS). Separation was performed according to the recently described LC-UV method of Ashenafi et al. (2011) [14]. This method was reported to have a good selectivity as it was able to separate the four main components of tylosin from the already known and 23 unknown impurities. However, as this method uses a mobile phase with non-volatile constituents, direct characterization of these impurities using LC/MS was not possible. The impurity fractions were therefore first collected and then desalted before sending them to the MS. Identification of the impurities in the tylosin sample was performed with a quadruple ion trap (IT) MS, with an electrospray ionization (ESI) source in the positive ion mode. The structure of the impurities was deduced by comparing their fragmentation pattern with those of the main components of tylosin. As several peaks in the LC-UV method contained multiple compounds, using this method in total 41 new impurities were (partly) characterized.