Merck
Semipermanent eyelash extensions causing bacterial keratitis: a case report.
Canadian journal of ophthalmology. Journal canadien d'ophtalmologie (2013-06-19)
Katja Ullrich, Niladri Saha
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
USP
Cefazolin, United States Pharmacopeia (USP) Reference Standard
Cefazolin, European Pharmacopoeia (EP) Reference Standard