Merck
  • Physical and chemical stability of ceftazidime and cefazolin in peritoneal dialysis solutions packaged in dual-chamber infusion bags.
Physical and chemical stability of ceftazidime and cefazolin in peritoneal dialysis solutions packaged in dual-chamber infusion bags.
American journal of health-system pharmacy : AJHP : official journal of the American Society of Health-System Pharmacists (2013-08-15)
Rahul P Patel, Siti Farawahida, Madhur Shastri, Troy Wanandy, Matthew D Jose
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
USP
Ceftazidim Pentahydrat, United States Pharmacopeia (USP) Reference Standard
USP
Cefazolin, United States Pharmacopeia (USP) Reference Standard
Ceftazidim, European Pharmacopoeia (EP) Reference Standard
Cefazolin, European Pharmacopoeia (EP) Reference Standard
Ceftazidim für die Peakidentifizierung, European Pharmacopoeia (EP) Reference Standard