Merck
  • [New and future treatments for neurological disorders--knowledge essential to daily clinics and future prospects. Topics: 9. Therapeutic progress in multiple sclerosis and neuromyelitis optica].
[New and future treatments for neurological disorders--knowledge essential to daily clinics and future prospects. Topics: 9. Therapeutic progress in multiple sclerosis and neuromyelitis optica].
Nihon Naika Gakkai zasshi. The Journal of the Japanese Society of Internal Medicine (2013-10-31)
Masami Tanaka, Keiko Tanaka
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
FTY720, ≥98% (HPLC)