Merck
  • Combination therapy of 15-epi-lipoxin A4 with antibiotics protects mice from Escherichia coli-induced sepsis*.
Combination therapy of 15-epi-lipoxin A4 with antibiotics protects mice from Escherichia coli-induced sepsis*.
Critical care medicine (2014-01-28)
Tomomi Ueda, Koichi Fukunaga, Hiroyuki Seki, Jun Miyata, Makoto Arita, Taku Miyasho, Toru Obata, Koichiro Asano, Tomoko Betsuyaku, Junzo Takeda
ZUSAMMENFASSUNG
Inflammation occurs along with infection during sepsis. 15-Epi-lipoxin A4 has protective and resolving effects in experimental models of infection. In this study, we examined the effects of 15-epi-lipoxin A4 combined with antibiotics on Escherichia coli-induced peritonitis. Prospective experimental study. University research laboratory. Male C57BL/6 mice. Mice were injected with E. coli to induce peritonitis and were given either 15-epi-lipoxin A4 (1 μg/mouse) or placebo (saline) with antibiotics (ceftazidime). The effects of 15-epi-lipoxin A4 on peritoneal cell populations, bacterial burden, and cytokine production were assessed. Survival rates were observed for up to 7 days. In addition, we examined the effects of 15-epi-lipoxin A4 on peritoneal macrophages stimulated with lipopolysaccharide, CpG DNA, or live E. coli. Treatment with 15-epi-lipoxin A4 significantly reduced the number of neutrophils in the peritoneum, inhibited production of cytokines and chemokines, and decreased bacterial load in the serum. Combined treatment of 15-epi-lipoxin A4 with antibiotics significantly improved survival in E. coli-infected mice. 15-Epi-lipoxin A4 also attenuated the production of interleukin-6 and tumor necrosis factor-α by lipopolysaccharide- or CpG DNA-stimulated peritoneal macrophages. Furthermore, 15-epi-lipoxin A4 combined with antibiotics synergistically reduced the production of interleukin-6 and tumor necrosis factor-α by peritoneal macrophages stimulated with live E. coli. 15-Epi-lipoxin A4 combined with antibiotics attenuated systemic inflammation, inhibited bacteria dissemination, and improved survival in E. coli-infected mice. The reduced production of interleukin-6 and tumor necrosis factor-α by peritoneal macrophages suggested that 15-epi-lipoxin A4 blocked the initial proinflammatory response. Taken together, these data suggested that 15-epi-lipoxin A4 combined with antibiotics was beneficial in regulating the proinflammatory response in sepsis without exacerbating infection.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
USP
Ceftazidim Pentahydrat, United States Pharmacopeia (USP) Reference Standard
Ceftazidim, European Pharmacopoeia (EP) Reference Standard
Ceftazidim für die Peakidentifizierung, European Pharmacopoeia (EP) Reference Standard