- [Pharmacological cardioversion of paroxysmal atrial fibrillation: 3 methods compared].
[Pharmacological cardioversion of paroxysmal atrial fibrillation: 3 methods compared].
La Clinica terapeutica (1987-07-31)
D Tsialtas, S Ghiringhelli, E Cozzi, F Glisenti, U Straneo, L Chittolini, G Straneo
PMID2973906
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Deslanosid, European Pharmacopoeia (EP) Reference Standard