Merck
  • Direct high-performance liquid chromatographic determination in urine of the enantiomers of propranolol and its major basic metabolite 4-hydroxypropranolol.
Direct high-performance liquid chromatographic determination in urine of the enantiomers of propranolol and its major basic metabolite 4-hydroxypropranolol.
Journal of chromatography (1993-02-26)
V L Herring, J A Johnson
ZUSAMMENFASSUNG
A method is described for quantitation of underivatized enantiomers of propranolol and its major basic metabolite, 4-hydroxypropranolol, in urine samples by high-performance liquid chromatography with fluorescence detection, using a cellulose tris(3,5-dimethylphenylcarbamate) chiral stationary phase. This method was found to be precise and accurate for the measurement of propranolol and 4-hydroxypropranolol concentrations in urine from pharmacokinetic investigations. This method represents the first assay for direct determination of 4-hydroxypropranolol enantiomers. The ability to easily measure 4-hydroxypropranolol enantiomers is valuable because the stereoselective disposition of propranolol is primarily due to stereoselective metabolism in the pathway responsible for generation of 4-hydroxypropranolol.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
(±)-4-Hydroxy-propranolol -hydrochlorid, analytical standard