Merck
  • The mu1 and mu2 opioid receptor binding of ketobemidone, norketobemidone and 3-dimethylamino-1,1-diphenylbutene.
The mu1 and mu2 opioid receptor binding of ketobemidone, norketobemidone and 3-dimethylamino-1,1-diphenylbutene.
Pharmacology & toxicology (1996-08-01)
K Kristensen, C B Christensen, L L Christrup, L C Nielsen
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
USP
Citalopram-verwandte Verbindung F, United States Pharmacopeia (USP) Reference Standard
Ketobemidon Unreinheit C, European Pharmacopoeia (EP) Reference Standard