Merck
  • Biosynthesis of gibberellic acid (GA3) and mycotoxins by F. moniliforme sheldon and other species of Liseola section.
Biosynthesis of gibberellic acid (GA3) and mycotoxins by F. moniliforme sheldon and other species of Liseola section.
Natural toxins (1996-01-01)
D Latus-Zietkiewicz, J Chelkowski, E Foremska, P Goliński, J Grabarkiewicz-Szczesna, M Kostecki, M Lew, J Perkowski, M Piasecki, M Wiewiórowska, K Szebiotko
ZUSAMMENFASSUNG
F. moniliforme and other species of Liseola section, F. culmorum, F. dlamini, and F. nygamai, were examined for their ability to produce gibberellic acid (GA3), fumonisins, trichothecenes, zearalenone, moniliformin, and bikaverin (TLC method). Gibberellic acid was produced by F. moniliforme strains in liquid medium and on rice kernels with a maximum concentration level of 470 mg/dm3 and 1 g/kg, respectively. No strain isolated in Poland produced GA3. High-yielding gibberellic acid strains produced neither trichothecenes and fumonisins nor other tested compounds. Also the rest of strains of examined species did not produce trichothecenes and other mycotoxins except for fumonisins which were found in rice cultures of F. moniliforme, F. proliferatum, and F. subglutinans. Bikaverin was produced by F. moniliforme always together with fumonisins. Filtrates of liquid cultures of gibberellin producing strains were tested for their toxicity to brine shrimps larvae (Artemia salina). It was found that GA3 presence does not increase toxicity of these filtrates.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Bikaverin, from Fusarium subglutinans, ≥98% (HPLC)