- Blood partition and protein binding of a new reversible proton pump inhibitor, YH1885.
Blood partition and protein binding of a new reversible proton pump inhibitor, YH1885.
Biopharmaceutics & drug disposition (1998-09-16)
K S Han, Y G Kim, J W Lee, M G Lee
PMID9737823
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Fenticonazolnitrat, European Pharmacopoeia (EP) Reference Standard