Merck
  • General synthesis of periodic macroporous solids by templated salt precipitation and chemical conversion.
General synthesis of periodic macroporous solids by templated salt precipitation and chemical conversion.
Chemistry of Materials (2000)