Merck
Serum autoantibody signature of ductal carcinoma in situ progression to invasive breast cancer.
Clinical cancer research : an official journal of the American Association for Cancer Research (2012-02-11)
Alain Mangé, Jérôme Lacombe, Caroline Bascoul-Mollevi, Marta Jarlier, Pierre-Jean Lamy, Philippe Rouanet, Thierry Maudelonde, Jérôme Solassol
ZUSAMMENFASSUNG
The identification of markers associated with progression to invasive breast cancer (IBC) is a major factor that can guide physicians in the initial therapeutic decision and the management of ductal carcinoma in situ (DCIS). We examined autoantibody targets in 20 DCIS and 20 IBC patients using protein microarrays and identified humoral responses that can be used to distinguish the two groups. The five most differentially targeted antigens were selected to generate an autoantibody signature for the in situ to invasive breast cancer transition. This signature was next tested on 120 independent samples (61 DCIS and 59 IBC) using specific ELISA assays. The prognosis value of the autoantibody signature was finally evaluated in a cohort of DCIS patients followed for 5 years. A set of five autoantibody targets (RBP-Jκ, HMGN1, PSRC1, CIRBP, and ECHDC1) with the highest differential signal intensity found in the protein microarrays experiment was used to establish an autoantibody signature of the DCIS to IBC transition. Using ELISA, this signature significantly discriminated DCIS from IBC [area under the ROC curve (AUC) = 0.794, 95% confidence interval (CI): 0.674-0.877]. Interestingly, our panel could highly distinguish low-grade DCIS from high-grade DCIS exhibiting an AUC of 0.749 (95% CI: 0.581-0.866). Finally, using a Kaplan-Meier analysis, the autoantibody signature could significantly divide the DCIS patients into a poor prognosis group and a good prognosis group (P = 0.01). These results indicate the potential of autoantibody detection as a new prognostic test with possible clinical implications for the management of DCIS.