Merck
  • Colorimetric 'naked-eye'and fluorescent sensors for anions based on amidourea functionalised 1, 8-naphthalimide structures: anion recognition via either deprotonation or hydrogen bonding in DMSO.
Colorimetric 'naked-eye'and fluorescent sensors for anions based on amidourea functionalised 1, 8-naphthalimide structures: anion recognition via either deprotonation or hydrogen bonding in DMSO.
New. J. Chem. (2008)
Ali HDP, et al.