Merck
  • A novel mechanism of alternative promoter and splicing regulates the epitope generation of tumor antigen CML66-L.
A novel mechanism of alternative promoter and splicing regulates the epitope generation of tumor antigen CML66-L.
Journal of immunology (Baltimore, Md. : 1950) (2003-12-23)
Yan Yan, Leuyen Phan, Fan Yang, Moshe Talpaz, Yu Yang, Zeyu Xiong, Bernard Ng, Nikolai A Timchenko, Catherine J Wu, Jerome Ritz, Hong Wang, Xiao-Feng Yang
ZUSAMMENFASSUNG
This report describes the difference in the epitope generation of two isoforms of self-tumor Ag CML66 and the regulation mechanism. We identified a new CML66 short isoform, termed CML66-S. The previously identified long CML66 is referred to as CML66-L. CML66-S shares the C terminus with CML66-L but has its unique N terminus. CML66-S is predominantly expressed in testis, but is also expressed in very low levels in tumor cells, whereas CML66-L is expressed in tumor cells and testis. Differential expression of CML66-L and CML66-S in tumor cells resulted from regulation at transcription, although alternative splicing also participated in the generation of the isoforms. In addition, Ab titers to a CML66-L peptide were significantly higher than that to CML66-S peptide in the sera from patients with tumors. Finally, the Abs to full-length CML66-L in the sera from patients with tumors were correlated with the Abs in the sera from these patients to CML66-L-38, which is a fusion protein with a CML66-L-specific N terminus. This suggests that the CML66-L isoform is mainly responsible for the epitope generation. Our studies have identified the alternative promoter in combination with alternative splicing as a novel mechanism for regulation of the epitope generation of a self-tumor Ag.